Tetrabromophthalic anhydride for synthesis, Sigma-Aldrich®
8.08096.0250
: Merck
:
- Pack type:Plastic bottle
- Pk:250 g
- Seq:3545
- Solubility:(20 °C) insoluble
- Melting Point:271 - 274 °C
- Molar Mass:463.7 g/mol
- Vapor Pressure:14.67 Pa (20 °C)
- Density:2.87 g/cm3 (20 °C)
:
:463.7 g/mol
:477 °C (1013 hPa)
:271…274 °C
:2,93 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
:211-185-4
:632-79-1
:C₈Br₄O₃
:MFCD00005919