3-Phenylpropanal for synthesis, Sigma-Aldrich®
Catalog # 8.04542.0100
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:100 ml
- Seq:2440
Specifications
Specification Test Results
- Solubility:0.74 mg/l (20 °C)
- Melting Point:-42 °C
- Molar Mass:134.18 g/mol
- Boiling Point:222 °C
- Vapor Pressure:15 hPa (98 °C)
- Flash Point:79 °C
- Density:1.01 g/cm3 (20 °C)
- Ignition Point:245 °C DIN 51794
Specification Test Results
About this item
:
:134.18 g/mol
Boiling Pt:224 °C (1013 hPa)
Melting Pt:–42 °C
Density:1,01 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:203-211-8
CAS Number:104-53-0
Formula:C₉H₁₀O
MDL Number:MFCD00007021