


Isobutyl chloroformate for synthesis, Sigma-Aldrich®
: Merck
:543-27-1
:
:C₅H₉ClO₂
:MFCD00000642
:136.58 g/mol
:13,05156
:128,7 °C (1013 hPa)
:–80 °C
:1,05 g/cm³ (20 °C)
:208-840-1
:2742
:6.1 (3, 8),II
:M-Clarity™ quality level = MQ200
:30 °C
:GHS02
:GHS06
:GHS05
:Ambient
- Solubility:(20 °C) (decomposition)
- Molar Mass:136.58 g/mol
- Boiling Point:127 - 129 °C (1013 hPa)
- Flash Point:30 °C
- Refractive Index:1.407 (20 °C, 589 nm)
- Density:1.05 g/cm3 (20 °C)
- Hazard Class:6.1
- Hazard Class Description:Toxic Materials
Frequently Bought Together