Order Entry
Ireland
ContactUsLinkComponent
Ethyl chloroformate for synthesis, Sigma-Aldrich®
Ethyl chloroformate for synthesis, Sigma-Aldrich®
Catalog # 8.00881.9025
Supplier:  Merck
CAS Number:  
Ethyl chloroformate for synthesis, Sigma-Aldrich®
Catalog # 8.00881.9025
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    25 L
  • Seq:
    5327

Specifications

About this item

 :
 :108.52 g/mol
Merck Index:12,03831
Boiling Pt:93 °C (1013 hPa)
Melting Pt:–81 °C
Density:1,14 g/cm³ (20 °C)
Flash Pt:10 °C
Storage Temperature:Ambient
EINECS:208-778-5
UN:1182
ADR:6.1 (3, 8),I
 :M-Clarity™ quality level = MQ200
 :GHS02
 :GHS06
 :GHS05
 :GHS09
CAS Number:541-41-3
Formula:C₃H₅ClO₂
MDL Number:MFCD00000644