

Ethyl methacrylate stabilised for synthesis, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:97-63-2
:
Formula:C₆H₁₀O₂
MDL Number:MFCD00009161
:114.14 g/mol
Boiling Pt:100 °C (1013 hPa)
Melting Pt:–48 °C
Density:0,9158 g/cm³ (20 °C)
Flash Pt:20 °C
UN:2277
ADR:3,II
:M-Clarity™ quality level = MQ200
EINECS:202-597-5
:GHS02
:GHS07
Specification Test Results
- Solubility:5.1 g/l (20 °C)
- Melting Point:-61 - -59 °C
- Molar Mass:114.14 g/mol
- Boiling Point:119 - 121 °C (1013 hPa)
- Vapor Pressure:21.3 hPa (20 °C)
- Flash Point:18 °C
- Refractive Index:1.4128 (20 °C, 589 nm)
- Explosion Limit:1.8 %(V)
- Density:0.913 g/cm3 (20 °C)
- Ignition Point:329 °C
Specification Test Results
Frequently Bought Together