


Nitric acid ≥69%, Puriss. p.a. ACS, ISO, Fluka™
Catalog # 30702-2.5L
Supplier: Honeywell Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:2,5 L
- Seq:5096
Specifications
Specification Test Results
- Assay:69 - 70 %
- Residue on ignition (SO₄):Max. 0.0005 %
- Silver (Ag):Max. 0.000001 %
- Aluminium (Al):Max. 0.000005 %
- Arsenic (As):Max. 0.000001 %
- Barium (Ba):Max. 0.000001 %
- Beryllium (Be):Max. 0.000001 %
- Bismuth (Bi):Max. 0.00001 %
- Calcium (Ca):Max. 0.00001 %
- Cadmium (Cd):Max. 0.000001 %
- Cobalt (Co):Max. 0.000001 %
- Chromium (Cr):Max. 0.000002 %
- Copper (Cu):Max. 0.000001 %
- Iron (Fe):Max. 0.00001 %
- Germanium (Ge):Max. 0.000005 %
- Potassium (K):Max. 0.00001 %
- Lithium (Li):Max. 0.000001 %
- Magnesium (Mg):Max. 0.00001 %
- Manganese (Mn):Max. 0.000001 %
- Molybdenum (Mo):Max. 0.000002 %
- Sodium (Na):Max. 0.00005 %
- Nickel (Ni):Max. 0.000002 %
- Lead (Pb):Max. 0.000001 %
- Strontium (Sr):Max. 0.000001 %
- Titanium (Ti):Max. 0.00001 %
- Thallium (Tl):Max. 0.000005 %
- Vanadium (V):Max. 0.000001 %
- Zinc (Zn):Max. 0.000005 %
- Zirconium (Zr):Max. 0.00001 %
- Heavy metals (as Pb):Max. 0.00002 %
- Chloride (Cl):Max. 0.00005 %
- Phosphate (PO₄):Max. 0.00005 %
- Sulfate (SO₄):Max. 0.00005 %
- APHA:Max. 10
Specification Test Results
About this item
:
:63.01 g/mol
Boiling Pt:122 °C (1,013 hPa)
Melting Pt:–38 °C
Density:1,420 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
ADR:8,II
:GHS03
:GHS05
:GHS06
CAS Number:7697-37-2
Formula:HNO₃
MDL Number:MFCD00011349