Human thrombin (from plasma)
Catalog # 605190-1000
Supplier: Merck Millipore (Calbiochem)
CAS Number:
Specifications
- Pk:1.000 EU
- CAS No.:9002-04-4
- Source:Plasma
- Species:Human
- Seq:02
- Enzyme name:Thrombin
- Enzyme activity:≥1000 NIH units/mg
Specifications
About this item
Key enzyme that activates factor XIII and converts fibrinogen into fibrin.