N-Phenyl-o-phenylenediamine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: N-(2-Aminophenyl)aniline, 2-Aminodiphenylamine, N1-Phenylbenzene-1,2-diamine
CAS Number:534-85-0
:
Formula:C₁₂H₁₂N₂
MDL Number:MFCD00007685
Boiling Pt:106…108 °C (0.4 mmHg)
Melting Pt:77…80 °C
Density:1.234 g/cm³ (20 °C)
Molecular Weight:184.24 g/mol
Storage Temperature:Ambient
EINECS:208-606-9
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together