Order Entry
Ireland
ContactUsLinkComponent
 
3-(4-Bromophenyl)propionic acid ≥98%
Catalog # H26785.06
CAS Number:  
3-(4-Bromophenyl)propionic acid ≥98%
Catalog # H26785.06
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :229.07 g/mol
Melting Pt:133…136 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:1643-30-7
Formula:C₉H₉BrO₂
MDL Number:MFCD01310793