4,5,6-Trifluoropyrimidine ≥97%
Catalog # H25855.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
CAS Number:17573-78-3
:
Formula:C₄HF₃N₂
:134.06 g/mol
Boiling Pt:83…85 °C
Density:1,4609
Flash Pt:>85 °C (185 °F)
Storage Temperature:Ambient
ADR:8,III
:GHS05
MDL Number:MFCD05662363