


N,N-Dimethylethylamine 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:598-56-1
:
Formula:C₄H₁₁N
MDL Number:MFCD00009039
:73.14 g/mol
Boiling Pt:36…37 °C
Melting Pt:–140 °C
Density:0,675
Flash Pt:–36 °C (–33 °F)
EINECS:209-940-8
ADR:3,II
:GHS02
:GHS05
:GHS07
Related Information
Frequently Bought Together