
Methyl-p-toluate ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:99-75-2
:
Formula:C₉H₁₀O₂
MDL Number:MFCD00008441
:150.18
Boiling Pt:221…224 °C
Melting Pt:32…35 °C
Density:1,060
Flash Pt:89 °C (192 °F)
EINECS:202-784-1
:GHS07
Storage Temperature:Ambient
Frequently Bought Together