

2-Mercaptobenzothiazole ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:149-30-4
:
Formula:C₇H₅NS₂
MDL Number:MFCD00005781
:167.26 g/mol
Merck Index:14,5868
Boiling Pt:302 °C (1013 hPa)
Melting Pt:178…182 °C
Density:1,420
Flash Pt:243 °C (469 °F)
Storage Temperature:Ambient
EINECS:205-736-8
UN:3077
ADR:9,III
:GHS07
:GHS09
Related Information
Frequently Bought Together