Sodium perborate tetrahydrate ≥97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: Sodium peroxometaborate tetrahydrate
CAS Number:13517-20-9
:10486-00-7
Formula:NaH₂BO₄·4H₂O
MDL Number:MFCD00149231
:171.87 g/mol
Merck Index:14,8652
Melting Pt:60 °C (dec.)
Density:1,73
EINECS:239-172-9
ADR:5.1,III
:GHS03
:GHS08
:GHS05
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Related Information
Frequently Bought Together