
1,3-Dioxolane ≥99.5% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:646-06-0
:
Formula:C₃H₆O₂
MDL Number:MFCD00003207
:74.08 g/mol
Boiling Pt:74…75 °C
Melting Pt:–95 °C
Density:1,06
Flash Pt:1 °C (34 °F)
UN:1166
ADR:3,II
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
EINECS:211-463-5
:GHS02
Storage Temperature:Ambient
Related Information
Frequently Bought Together