

tert-Butyl ethyl ether 97%
:637-92-3
:
:C₆H₁₄O
:MFCD00009225
:102.18 g/mol
:13,03809
:71 °C (1013 hPa)
:–94 °C
:0,75 g/cm³
:–19 °C
:Ambient
:211-309-7
:1179
:3,II
:GHS02
:GHS07
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.3746 to 1.3766 (20°C, 589 nm)
- GC:≥96.0 %
Frequently Bought Together