Order Entry
Ireland
ContactUsLinkComponent
3-Fluoropyrocatechol 98+%
3-Fluoropyrocatechol 98+%
Certificates
undefined
3-Fluoropyrocatechol 98+%

Specification Test Results

  • Appearance:
    Light beige to grey-brown Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    69 to 73 °C
  • GC:
    ≥98.0 %

Specification Test Results

Related Information

Frequently Bought Together