

Ethyl methacrylate 99% stabilised
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:97-63-2
:
Formula:C₆H₁₀O₂
MDL Number:MFCD00009161
:114.14 g/mol
Boiling Pt:118…119 °C (760 mmHg)
Melting Pt:–75 °C
Density:0,91 g/cm³
Flash Pt:15 °C
Storage Temperature:Refrigerator
EINECS:202-597-5
UN:2277
ADR:3,II
:GHS07
:GHS02
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4120 to 1.4160 (20°C, 589 nm)
- GC:≥98.5 %
- Stabilizer:10 to 120 ppm MEHQ
Specification Test Results
Frequently Bought Together