
meso-Tetraphenylporphine 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:917-23-7
:
Formula:C₄₄H₃₀N₄
MDL Number:MFCD00011680
:614.75 g/mol
Melting Pt:>300 °C
Density:1,276 g/cm³ (20 °C)
EINECS:213-025-9
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Blue to purple Crystalline powder or crystals
- Infrared spectrum:Conforms
- HPLC:≥96.0 %
- Impurity:≤3 % Chlorin
Specification Test Results
Frequently Bought Together