




1-(Trimethylsilyl)imidazole 97%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:18156-74-6
:
Formula:C₆H₁₂N₂Si
MDL Number:MFCD00005280
:140.26 g/mol
Boiling Pt:93…94 °C (13 mmHg)
Melting Pt:–42 °C
Density:0,95 g/cm³
Flash Pt:5 °C
EINECS:242-040-3
UN:2733
ADR:3,II
:GHS02
:GHS05
:GHS07
:GHS08
:GHS09
Storage Temperature:Refrigerator
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4750 to 1.4780 (20°C, 589 nm)
- GC:≥96.0 %
- Insoluble matter:≤0.01 % (10 % in water)
Specification Test Results
Frequently Bought Together