


N,O-Bis(trimethylsilyl)acetamide 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:10416-59-8
:
Formula:C₈H₂₁NOSi₂
MDL Number:MFCD00008270
:203.43 g/mol
Boiling Pt:71…73 °C (35 mmHg)
Melting Pt:–8 °C
Density:0,82 g/cm³
Flash Pt:42 °C
Storage Temperature:Refrigerator
EINECS:233-892-7
UN:2920
ADR:8,II
:GHS07
:GHS05
:GHS02
Specification Test Results
- Appearance:Clear colorless to yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4160 to 1.4190 (20°C, 589 nm)
- GC:≥94 %
Specification Test Results
Frequently Bought Together