

Octadecyltrichlorosilane 95%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:112-04-9
:
Formula:C₁₈H₃₇Cl₃Si
MDL Number:MFCD00000484
:387.94 g/mol
Boiling Pt:223 °C (10 mmHg)
Melting Pt:22 °C
Density:0,98 g/cm³
Flash Pt:189 °C
Storage Temperature:Ambient
EINECS:203-930-7
UN:1800
ADR:8,II
:GHS05
:GHS07
Specification Test Results
- Appearance:Clear colorless to yellow-brown Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4585 to 1.4605 (20°C, 589 nm)
- GC:≥94 %
Specification Test Results
Frequently Bought Together