
3,4-Dimethoxybenzylamine 97%
:5763-61-1
:
:C₉H₁₃NO₂
:MFCD00008116
:167.21
:281…284 °C (760 mmHg)
:1,1 g/cm³
:>112 °C
:Ambient
:227-287-7
:2735
:8,II
:GHS05
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5530 to 1.5570 (20°C, 589 nm)
- GC:≥96.0 %
Frequently Bought Together