Order Entry
Ireland
ContactUsLinkComponent
 
Organic reference standard, Styrene 200 µg/ml in methanol
Catalog # ACCUM-502-42
Supplier:  AccuStandard
CAS Number:  
Organic reference standard, Styrene 200 µg/ml in methanol
Catalog # ACCUM-502-42
Supplier:  AccuStandard
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL
  • CAS No.:
    100-42-5
  • Description:
    Styrene 200 µg/ml in methanol
  • Formula:
    C₈H₈ 
  • Description:
    Styrene

Specifications

About this item