Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Chlorobenzene standard 200 µg/ml in methanol
Chlorobenzene standard 200 µg/ml in methanol
Chlorobenzene standard 200 µg/ml in methanol
Catalog # ACCUM-502-11
Supplier:
AccuStandard
CAS Number:
Expand image
Chlorobenzene standard 200 µg/ml in methanol
Catalog # ACCUM-502-11
Supplier:
AccuStandard
CAS Number:
Specifications
Pk:
1 mL
CAS No.:
108-90-7
Description:
Chlorobenzene
Formula:
C₆H₅Cl
Description:
Chlorobenzene
Specifications
About this item
Product Details
Product Details