

Uridine-5'-triphosphoric acid trisodium salt (UTP-Na3), white ≥97%, powder
Catalog # 02EFC05591
Supplier: MP Biomedicals
CAS Number:
Specifications
- Pk:1 kg
- Seq:4024
Specifications
About this item
:
:550.09 g/mol
ADR:H302-H371,
:GHS07
:GHS08
CAS Number:19817-92-6
Formula:C₉H₁₂N₂Na₃O₁₅P₃
MDL Number:MFCD00044310