
Methyl 3-chloro-6-methoxy-1-benzothiophene-2-carboxylate 95%
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:59812-35-0
:
Formula:C₁₁H₉ClO₃S
MDL Number:MFCD00620636
:256.71 g/mol
:GHS07
Storage Temperature:Refrigerator
Frequently Bought Together