Order Entry
Ireland
ContactUsLinkComponent
1-Boc-indole-2-boronic acid 98%
Irritant
1-Boc-indole-2-boronic acid 98%
Catalog # APOSOR10393-500MG
Supplier:  Apollo Scientific
CAS Number:  
Irritant
1-Boc-indole-2-boronic acid 98%
Catalog # APOSOR10393-500MG
Supplier:  Apollo Scientific
CAS Number:  

Specifications

  • Pack type:
    Bottle
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :261.08 g/mol
Melting Pt:85 - 90 °C
 :GHS07
Storage Temperature:Freezer
CAS Number:213318-44-6
Formula:C₁₃H₁₆BNO₄
MDL Number:MFCD02093045
Melting Pt:85…90 °C