
2-Bromo-3-fluoro-isonicotinonitrile 95%
:1805188-90-2
:
:C₆H₂BrFN₂
:MFCD28735371
:201 g/mol
:GHS07
:Refrigerator
Frequently Bought Together
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for UHPLC-MS
: VWR Chemicals
Dichloromethane, anhydrous ≥99.7% stabilised
: Thermo Fisher Scientific
N,N-Dimethylformamide, Reagent Grade
: VWR Chemicals
Ethyl acetate ≥99.9%, CHROMASOLV™ for pesticide residue analysis, Riedel-de Haën™
: Honeywell Chemicals
2,2,4-Trimethylpentane ≥99.7% for HPLC
: Thermo Fisher Scientific
2-Propanol ≥99.9%, CHROMASOLV™ for LC-MS, Riedel-de Haën™
: Honeywell Chemicals
Water, Ultrapure for HPLC
: Thermo Fisher Scientific