2,4,6-Trimethyl-m-phenylenediamine
Supplier: Apollo Scientific
Certificates
About this item
CAS Number:3102-70-3
:
Formula:C₉H₁₄N₂
MDL Number:MFCD00010149
:150.22 g/mol
Melting Pt:89…91 °C
UN:3077
ADR:9+EHS,III
:GHS07
Frequently Bought Together