Supelco® Organic Reference Standard, 2,4,6-Trichloroanisole solution
Catalog # 47526-U
Supplier: Merck
CAS Number:
Specifications
- Concentration:100 μg/ml in methanol
- Pk:1
- Standards:Certified reference material
- CAS No.:87-40-1
- Description:2,4,6-Trichloroanisole solution
- Storage conditions:2...30 °C
- Hazard class description:H225 - H301 + H311 + H331 - H370
- Shipping conditions:STD
- M-Clarity quality level:MQ100
- Flash point:11 °C
Specifications
About this item
- GHS signal: GHS02, GHS06, GHS08
Caution: P210 - P233 - P280 - P301 + P310 - P303 + P361 + P353 - P304 + P340 + P311