Order Entry
Ireland
ContactUsLinkComponent
 
Organic reference standard, 1-Bromo-4-fluorobenzene 2000 µg/ml in methanol, Supelco®
Catalog # 48083.
Supplier:  Merck
CAS Number:  
Organic reference standard, 1-Bromo-4-fluorobenzene 2000 µg/ml in methanol, Supelco®
Catalog # 48083.
Supplier:  Merck
CAS Number:  

Specifications

  • Pk:
    1
  • CAS No.:
    460-00-4
  • Description:
    1-Bromo-4-fluorobenzene 2,000 µg/ml in methanol
  • Formula:
    C₆H₄BrF
  • Description:
    1-Bromo-4-fluorobenzene

Specifications

About this item