Order Entry
Ireland
ContactUsLinkComponent
Lecithin from egg yolk
Lecithin from egg yolk
Catalog # ROTH7514.1
Supplier:  Roth Carl
CAS Number:  
Lecithin from egg yolk
Catalog # ROTH7514.1
Supplier:  Roth Carl
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g
  • Seq:
    3308

Specifications

About this item

 :
EINECS:297-639-2
CAS Number:93685-90-6
MDL Number:MFCD00131449