Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cy3 tetrazine
Cy3 tetrazine fluorescent probe
Cy3 tetrazine fluorescent probe
Cy3 tetrazine fluorescent probe
AATB910
:
AAT Bioquest
:
Cy3 tetrazine fluorescent probe
AATB910
:
AAT Bioquest
:
Pack type:
Plastic bottle
Pk:
1 mg
Seq:
2070
:
:
908.07 g/mol
:
5,II
:
GHS07