0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Ireland
ContactUsLinkComponent
FMOC-Asp(5-TAMRA)-OH
FMOC-Asp(5-TAMRA)-OH for biomolecular labeling
FMOC-Asp(5-TAMRA)-OH for biomolecular labeling
Irritant
FMOC-Asp(5-TAMRA)-OH for biomolecular labeling
Catalog # AATB5006
Supplier:
AAT Bioquest
CAS Number:
Expand image
Irritant
FMOC-Asp(5-TAMRA)-OH for biomolecular labeling
Catalog # AATB5006
Supplier:
AAT Bioquest
CAS Number:
Specifications
Pack type:
Plastic bottle
Pk:
100 mg
Seq:
2440
Specifications
About this item
Product Details
:
ADR:
5,II
:
GHS07
Product Details