Order Entry
Ireland
ContactUsLinkComponent
Propargyl-PEG1-NHS ester for biomolecular labeling
Irritant
Propargyl-PEG1-NHS ester for biomolecular labeling
Catalog # AATB4531
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
Propargyl-PEG1-NHS ester for biomolecular labeling
Catalog # AATB4531
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
ADR:5,II
 :225.2 g/mol
 :GHS07
CAS Number:1174157-65-3
Formula:C₁₀H₁₁NO₅
MDL Number:MFCD13184973