Order Entry
Ireland
ContactUsLinkComponent
2-NBDG ≥90%
Irritant
2-NBDG ≥90%
Catalog # AATB36702
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
2-NBDG ≥90%
Catalog # AATB36702
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :342.3 g/mol
ADR:5,II
 :GHS07
CAS Number:186689-07-6
Formula:C₁₂H₁₄N₄O₈
MDL Number:MFCD23135882