
Tide Fluor™ 4, succinimidyl ester, Tide Fluor™ fluorescent probe
Catalog # AATB2289
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:5 mg
- Seq:2210
Specifications
About this item
:
:659.73 g/mol
ADR:5,II
:GHS07