Order Entry
Ireland
ContactUsLinkComponent
Tide Quencher™ 7WS maleimide, Tide Quencher™ fluorescent probe
Irritant
Tide Quencher™ 7WS maleimide, Tide Quencher™ fluorescent probe
Catalog # AATB2109
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
Tide Quencher™ 7WS maleimide, Tide Quencher™ fluorescent probe
Catalog # AATB2109
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
ADR:5,II
 :1109.24 g/mol
 :GHS07