Order Entry
Ireland
ContactUsLinkComponent
6-Carboxy-4',5'-dichloro-2',7'-dimethoxyfluorescein, succinimidyl ester ≥90%
Irritant
6-Carboxy-4',5'-dichloro-2',7'-dimethoxyfluorescein, succinimidyl ester ≥90%
Catalog # AATB203
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
6-Carboxy-4',5'-dichloro-2',7'-dimethoxyfluorescein, succinimidyl ester ≥90%
Catalog # AATB203
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :602.34 g/mol
ADR:5,II
 :GHS07
CAS Number:113394-23-3
Formula:C₂₇H₁₇Cl₂NO₁₁