Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
DNP-PEG4-acid
DNP-PEG4-acid FRET quencher precursor
DNP-PEG4-acid FRET quencher precursor
AATB2022
:
AAT BIOQUEST
:
DNP-PEG4-acid FRET quencher precursor
AATB2022
:
AAT BIOQUEST
:
Pack type:
Plastic bottle
Pk:
25 mg
Seq:
2315
:
:
431.395 g/mol
:
5,II
:
GHS07
:
858126-76-8
:
C₁₇H₂₅N₃O₁₀
:
MFCD13184986