Order Entry
Ireland
ContactUsLinkComponent
Biotin-16-dUTP 1 mM in TRIS buffer ≥95% pH 7.5
Irritant
Biotin-16-dUTP 1 mM in TRIS buffer ≥95% pH 7.5
Catalog # AATB17017
Supplier:  AAT BIOQUEST
CAS Number:  
undefined
Irritant
Biotin-16-dUTP 1 mM in TRIS buffer ≥95% pH 7.5
Catalog # AATB17017
Supplier:  AAT BIOQUEST
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    25 Nmo
  • Seq:
    0311

Specifications

About this item

 :
 :1013.72 g/mol
ADR:5,II
 :GHS07
CAS Number:136632-31-0