Order Entry
Ireland
ContactUsLinkComponent
Cyanine 7 azide ≥95% [equivalent to Cy7® azide]
Cyanine 7 azide ≥95% [equivalent to Cy7® azide]
Catalog # AATB163
Supplier:  AAT Bioquest
CAS Number:  
undefined
Cyanine 7 azide ≥95% [equivalent to Cy7® azide]
Catalog # AATB163
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

About this item

 :
 :864.95 g/mol
CAS Number:1557149-65-1
 :1557397-59-7