Order Entry
Ireland
ContactUsLinkComponent
Cbz-IETD-AFC ≥90%
Irritant
Cbz-IETD-AFC ≥90%
Catalog # AATB13425
Supplier:  AAT Bioquest
CAS Number:  
undefined
Irritant
Cbz-IETD-AFC ≥90%
Catalog # AATB13425
Supplier:  AAT Bioquest
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    5 mg
  • Seq:
    2210

Specifications

About this item

 :
 :821.76 g/mol
ADR:5,II
 :GHS07
CAS Number:219138-02-0
Formula:C₃₇H₄₂F₃N₅O₁₃
MDL Number:MFCD01318832