Native 2',3'-cGAMP (from Mammalian cells), Cy5®
Catalog # AATB20318
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Vial
- Pk:100 µG
- Conjugation:Cy5®
- Protein/peptide type:Native
- Source:Mammalian cells
- Protein synonyms:Cyclic guanosine monophosphate"adenosine monophosphate, cGAMP
- Protein/peptide name:2',3'-cGAMP
Specifications
About this item
2',3'-cGAMP or cGAMP (Cyclic guanosine monophosphate"adenosine monophosphate) is produced in mammalian cells by cGAS (cGAMP synthase) from ATP and GTP upon cytosolic DNA stimulation.
- Essential component of intracellular signaling
Caution: For research use only.