Order Entry
Ireland
ContactUsLinkComponent
Human ERBB2
Human ERBB2
Catalog # ORIGTS412583P5
Supplier:  OriGene
CAS Number:  
Human ERBB2
Catalog # ORIGTS412583P5
Supplier:  OriGene
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Species:
    Human
  • Tag sequence:
    C-MYC/DDK
  • Storage conditions:
    RT
  • Protein synonyms:
    NEU|TKR1|MLN 19|CD340|HER2|HER-2/neu|HER-2|NGL
  • Accession no.:
    NM_004448, NP_004439
  • Protein/peptide name:
    ERBB2
  • Pk Info:
    25 slides

Specifications

About this item

HEK293T cells were transiently transfected with TrueORF cDNA plasmid. Transfected cells were cultured for 48 hrs. After harvesting, the cultured cells were fixed in formalin and dehydrated before embedding in paraffin. 5 µm sections of the FFPE cell pellet blocks are cut and mounted on positively charged SuperFrost slides.

  • Expression host: HEK293T

Stability: Slides are guaranteed for a year from the date of receipt if proper storage instructions were followed

Caution: This product is for research use only and is not approved for use in humans or in clinical diagnosis.