Recombinant DLDVPIPGRFDRRVpSVAAE
Catalog # AATB31700
Supplier: AAT Bioquest
CAS Number:
Specifications
- Pack type:Vial
- Pk:1 mg
- Conjugation:Unconjugated
- Protein/peptide type:Recombinant
- Protein synonyms:H-Asp-Leu-Asp-Val-Pro-Ile-Pro-Gly-Arg-Phe-Asp-Arg-Arg-Val-Ser(PO₃H₂)-Val-Ala-Ala-Glu-OH
- Protein/peptide name:DLDVPIPGRFDRRVpSVAAE
- Molecular weight:2192,34
Specifications
About this item
DLDVPIPGRFDRRVpSVAAE is an excellent peptide substrate for calcineurin (Protein Phosphatase 2B, PP2B).
- Derived from the PKA regulatory subunit type II (RII)
The sequence is derived from the PKA regulatory subunit type II (RII) and is phosphorylated on the Ser residue.
Caution: For research use only.