Native Dextran (from Eukaryotic cells), Rhodamine 2
AATB20451
:
- Pack type:Vial
- Pk:1 mg
- Conjugation:Rhodamine 2
- Protein/peptide type:Native
- Source:Eukaryotic cells
- Protein synonyms:Manninotriose|Macrodex
- Protein/peptide name:Dextran
Dextran-conjugated calcium indicators are stably retained within neurons.
- Well-retained in neurons
- 10000 mw
- Useful for in vivo imaging
: For research use only.