Amplite® Fluorimetric Proteasome 20S Activity Assay Kit *Green Fluorescence*
AATB13456
:
- Pk:100 Tests
- Description:Amplite™ Fluorimetric proteasome 20S activity assay kit *green fluorescence*
- Tests per kit:100 Tests
Our Amplite® Fluorometric 20S Proteasome Assay Kit is a homogeneous fluorescent assay that measures the chymotrypsin-like protease activity associated with the proteasome complex either in cultured cells or cell lysates.
- In-house QC testing
- Sensitive
- Suggested protocol available
: For research use only.