Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Ireland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1X4 Color™ B488 Dye Set
1X4 Color™ B488 Dye Set
1X4 Color™ B488 Dye Set
AATB8005
:
AAT Bioquest
:
1X4 Color™ B488 Dye Set
AATB8005
:
AAT Bioquest
:
Pack type:
Plastic bottle
Pk:
1 SET
Seq:
0125
:
:
5,II
:
GHS07